missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-4782012
32.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13477-91-3
Nd2(SO4)3·8H2O
MFCD00149803
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13477-91-3
Nd2(SO4)3·8H2O
MFCD00149803

It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1176°C
Color Pink
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.9%
Solubility Information Soluble in water
Formula Weight 720.78 (576.66 Anhydrous)
Odor Odorless
Appearance Pink crystals
Density 2.85 g/cm3 at 20°C
Chemical Name or Material Neodymium(III) sulfate octahydrate, Rare Earth Oxides Content
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'