missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) isopropoxide, 99% (REO), 2.5% w/v in toluene/isopropanol, Thermo Scientific Chemicals

Catalog Number p-7050349
64.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19236-15-8
C9H24NdO3
324.53
MFCD00058819,MFCD00058819
WOCLHTSADRFWMF-UHFFFAOYSA-N
neodymium i-propoxide, neodymium iii isopropoxide
25199945
[Nd].CC(C)O.CC(C)O.CC(C)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19236-15-8
C9H24NdO3
324.53
MFCD00058819,MFCD00058819
WOCLHTSADRFWMF-UHFFFAOYSA-N
neodymium i-propoxide, neodymium iii isopropoxide
25199945
[Nd].CC(C)O.CC(C)O.CC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue
Linear Formula Nd[OCH(CH3)2]3
Quantity 25 mL
UN Number UN1993
Sensitivity Moisture sensitive
IUPAC Name tris(propan-2-ol) neodymium
Formula Weight 321.3
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 2.5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Neodymium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'