Learn More
Alfa Aesar™ Neodymium(III) bromide, ultra dry, 99.99% (metals basis)
Brand: Alfa Aesar™ 043672.22
Code : NEW
Additional Details : CAS Number : 13536-80-6 Weight : 0.10000kg
Quantity | 100g |
---|
Chemical Identifiers
Neodymium (III) bromide | |
Ultra dry | |
383.97 |
13536-80-6 | |
NdBr3 | |
MFCD00016245 |
Specifications
Neodymium (III) bromide | |
13536-80-6 | |
99.99% | |
NdBr3 | |
MFCD00016245 | |
Ampule | |
1540°C | |
100g |
Ultra dry | |
Beads | |
(metals basis) | |
383.97 | |
−10 Mesh | |
Hygroscopic | |
684°C |