missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nemonapride, Tocris Bioscience™
Highly potent D2-like antagonist. Also 5-HT1A agonist
Brand: Tocris (Bio-Techne) 174650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 50mg |
---|
Description
Highly potent dopamine D2-like receptor antagonist; selective over D1-like receptors (Ki values are 0.1 and 740 nM for D2-like and D1-like receptors respectively). Also potent 5-HT1A receptor agonist (IC50 = 34 nM) and has affinity for sigma receptors.Chemical Identifiers
75272-39-8 | |
387.908 | |
nemonapride, --nemonapride, 2s,3s-nemonapride, emonapride, emilace, n-2s,3s-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylamino benzamide, dsstox_cid_22612, dsstox_rid_80057, dsstox_gsid_42612, cis-n-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylaminobenzamide | |
CHEBI:64218 | |
CC1C(CCN1CC2=CC=CC=C2)NC(=O)C3=CC(=C(C=C3OC)NC)Cl |
C21H26ClN3O2 | |
KRVOJOCLBAAKSJ-KSSFIOAISA-N | |
9952220 | |
N-[(2S,3S)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide |
Specifications
Nemonapride | |
75272-39-8 | |
nemonapride, --nemonapride, 2s,3s-nemonapride, emonapride, emilace, n-2s,3s-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylamino benzamide, dsstox_cid_22612, dsstox_rid_80057, dsstox_gsid_42612, cis-n-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylaminobenzamide | |
CC1C(CCN1CC2=CC=CC=C2)NC(=O)C3=CC(=C(C=C3OC)NC)Cl | |
387.908 | |
CHEBI:64218 | |
>99% |
50mg | |
C21H26ClN3O2 | |
KRVOJOCLBAAKSJ-KSSFIOAISA-N | |
N-[(2S,3S)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide | |
9952220 | |
387.91 |