missing translation for 'onlineSavingsMsg'
Learn More

Nemonapride, Tocris Bioscience™

Highly potent D2-like antagonist. Also 5-HT1A agonist

Brand:  Tocris (Bio-Techne) 174610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448693

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent dopamine D2-like receptor antagonist; selective over D1-like receptors (Ki values are 0.1 and 740 nM for D2-like and D1-like receptors respectively). Also potent 5-HT1A receptor agonist (IC50 = 34 nM) and has affinity for sigma receptors.
Specifications

Chemical Identifiers

75272-39-8
387.908
nemonapride, --nemonapride, 2s,3s-nemonapride, emonapride, emilace, n-2s,3s-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylamino benzamide, dsstox_cid_22612, dsstox_rid_80057, dsstox_gsid_42612, cis-n-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylaminobenzamide
CHEBI:64218
CC1C(CCN1CC2=CC=CC=C2)NC(=O)C3=CC(=C(C=C3OC)NC)Cl
C21H26ClN3O2
KRVOJOCLBAAKSJ-KSSFIOAISA-N
9952220
N-[(2S,3S)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide

Specifications

10mg
75272-39-8
nemonapride, --nemonapride, 2s,3s-nemonapride, emonapride, emilace, n-2s,3s-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylamino benzamide, dsstox_cid_22612, dsstox_rid_80057, dsstox_gsid_42612, cis-n-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylaminobenzamide
CC1C(CCN1CC2=CC=CC=C2)NC(=O)C3=CC(=C(C=C3OC)NC)Cl
387.908
CHEBI:64218
>99%
Nemonapride
C21H26ClN3O2
KRVOJOCLBAAKSJ-KSSFIOAISA-N
N-[(2S,3S)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide
9952220
387.91
Videos
Documents