missing translation for 'onlineSavingsMsg'
Learn More

Nelfinavir mesylate, Tocris Bioscience™

Potent HIV-1 protease inhibitor

Brand:  Tocris (Bio-Techne) 376650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13463574

  • £445.92 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Orally active human immunodeficiency virus protease inhibitor. Potently inhibits HIV-1 protease (Ki = 2 nM) in vitro.
Specifications

Chemical Identifiers

159989-65-8
663.889
nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777
CHEBI:7497
CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O
C33H49N3O7S2
NQHXCOAXSHGTIA-SKXNDZRYSA-N
64142
(3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid

Specifications

Nelfinavir mesylate
159989-65-8
nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777
CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O
663.889
CHEBI:7497
>98%
50mg
C33H49N3O7S2
NQHXCOAXSHGTIA-SKXNDZRYSA-N
(3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid
64142
663.89
Videos