Learn More
Epredia™ Neg-50™ Frozen Section Medium
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
Brand: Epredia™ 6502Y
Code : LE
Additional Details : CAS Number : 7732-18-5 Weight : 0.36000kg
Description
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
- Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
- Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Specifications
Yellow | |
2/Cs. | |
4 oz. per bottle |