missing translation for 'onlineSavingsMsg'
Learn More

Nefazodone hydrochloride, Tocris Bioscience™

5-HT2A antagonist and 5-HT uptake inhibitor. Antidepressant

Brand:  Tocris (Bio-Techne) 277750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13460574

  • £461.52 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Serotonin 5-HT2A receptor antagonist (Ki = 5.8 nM) and inhibitor of serotonin and noradrenalin uptake (IC50 values are 290 and 300 nM respectively). Displays no activity at 5-HT1B and 5-HT1D receptors. Active in models predictive of antidepressant potential.
Specifications

Chemical Identifiers

82752-99-6
506.472
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, nefazodone hydrochloride, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
CHEBI:7495
CCC1=NN(C(=O)N1CCOC2=CC=CC=C2)CCCN3CCN(CC3)C4=CC(=CC=C4)Cl.Cl
C25H33Cl2N5O2
DYCKFEBIOUQECE-UHFFFAOYSA-N
54911
2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride

Specifications

Nefazodone hydrochloride
82752-99-6
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, nefazodone hydrochloride, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
CCC1=NN(C(=O)N1CCOC2=CC=CC=C2)CCCN3CCN(CC3)C4=CC(=CC=C4)Cl.Cl
506.472
CHEBI:7495
>99%
50mg
C25H33Cl2N5O2
DYCKFEBIOUQECE-UHFFFAOYSA-N
2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride
54911
506.47
Videos