missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nectin-4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Nectin-4/PVRL4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Nectin-4/PVRL4 Polyclonal specifically detects Nectin-4/PVRL4 in Mouse samples. It is validated for Western Blot.Specifications
Nectin-4/PVRL4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4 | |
PVRL4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q8R007 | |
81607 | |
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-4 Tyr28-Ile349 Accession # Q8R007 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Nectin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Nectin-4 is observed and less than 5% cross-reactivity with recombinant mouse Nectin-1, -2, and -3 is observed. |
For Research Use Only