missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-3 Mouse anti-Human, Clone: 337407, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Nectin-3/PVRL3 |
---|---|
Clone | 337407 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Nectin-3 Leu56-Asp400 Accession # Q9NQS3 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
Nectin-3/PVRL3 Monoclonal specifically detects Nectin-3/PVRL3 in Human samples. It is validated for Western Blot.Specifications
Nectin-3/PVRL3 | |
Mouse myeloma cell line NS0-derived recombinant human Nectin-3 Leu56-Asp400 Accession # Q9NQS3 | |
Primary | |
Human | |
Unconjugated | |
25945 | |
Purified | |
Western Blot | |
RUO |
337407 | |
Mouse | |
Monoclonal | |
IgG1 | |
CD113, CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, Nectin3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRL3, PVRR3 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only