missing translation for 'onlineSavingsMsg'
Learn More

Statebourne Cryogenics™ Neck Plugs

Brand:  Statebourne Cryogenics™ 9902055

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11909394

  • £107.85 / Each


Estimated Shipment
19-02-2018
Add to basket

Description

Description

  • Durable aluminium construction with super insulated high vacuum system to ensure high performance
  • Liquid nitrogen refrigerators with capacities from 2L to 36L offering a choice of holding times and specimen capacity
  • Lockable lids in high strength polycarbonate
  • CE marked under the Medical Devices Directive
  • Supplied with canisters and neckplug
Specifications

Specifications

Neck Plug
8, 10, 20 & 35L Refrigerator.
Videos
Documentation

Technical Documents