missing translation for 'onlineSavingsMsg'
Learn More

NE 100 hydrochloride, Tocris Bioscience™

Selective σ1 antagonist

Brand:  Tocris (Bio-Techne) 313350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13432494

  • £676.20 / 50mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective σ1 receptor antagonist (Ki = 0.86 nM) that displays > 55-fold selectivity over σ2 receptors and > 6000-fold selectivity over D1, D2, 5-HT1A, 5-HT2 and PCP receptors. Exhibits reversible binding (Kd = 1.2 nM) and displays antipsychotic activity in vivo. Orally active.
Specifications

Chemical Identifiers

149409-57-4
391.98
ne 100 hydrochloride, 4-methoxy-3-2-phenylethoxy-n,n-dipropylbenzeneethanamine hydrochloride, benzeneethanamine, 4-methoxy-3-2-phenylethoxy-n,n-dipropyl-, hydrochloride, c23h33no2.hcl
N-[2-[4-methoxy-3-(2-phenylethoxy)phenyl]ethyl]-N-propylpropan-1-amine;hydrochloride
C23H34ClNO2
ZHGMDXSHODHWHV-UHFFFAOYSA-N
9952420
CCCN(CCC)CCC1=CC(=C(C=C1)OC)OCCC2=CC=CC=C2.Cl

Specifications

50mg
149409-57-4
ne 100 hydrochloride, 4-methoxy-3-2-phenylethoxy-n,n-dipropylbenzeneethanamine hydrochloride, benzeneethanamine, 4-methoxy-3-2-phenylethoxy-n,n-dipropyl-, hydrochloride, c23h33no2.hcl
CCCN(CCC)CCC1=CC(=C(C=C1)OC)OCCC2=CC=CC=C2.Cl
391.98
391.97
NE 100 hydrochloride
C23H34ClNO2
ZHGMDXSHODHWHV-UHFFFAOYSA-N
N-[2-[4-methoxy-3-(2-phenylethoxy)phenyl]ethyl]-N-propylpropan-1-amine;hydrochloride
9952420
>98%
Videos
Documents