Learn More
NDUFAF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561446
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LAQVKDSVSE KTIGLMRMQF WKKTVEDIYC DNPPHQPVAI ELWKAVKRHN LTKRWLMKIV DEREKNLDDK AYRNIKELEN YA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.
NDUFAF6 Polyclonal antibody specifically detects NDUFAF6 in Human samples. It is validated for Immunohistochemistry.Specifications
NDUFAF6 | |
Polyclonal | |
Unconjugated | |
NDUFAF6 | |
C8orf38; NADH dehydrogenase (ubiquinone) complex I, assembly factor 6; NADH:ubiquinone oxidoreductase complex assembly factor 6; NDUFAF6; Putative phytoene synthase; UPF0551 protein C8orf38, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
137682 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q330K2 | |
NDUFAF6 | |
Recombinant protein corresponding to Human NDUFAF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only