missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ndelta-Fmoc-L-ornithine, 95%

Brand:  Alfa Aesar™ H63937.03

Code : NEW

Additional Details : CAS Number : 147071-84-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15470919

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Ndelta-Fmoc-L-ornithine
354.4
H-Orn(Fmoc)-OH
147071-84-9
MFCD00237208

Specifications

Ndelta-Fmoc-L-ornithine
Solid
354.4
H-Orn(Fmoc)-OH
147071-84-9
95%
MFCD00237208
1g
Videos