missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals

Catalog Number 15499278
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
109425-55-0
C25H30N2O6
454.523
MFCD00065668
JOOIZTMAHNLNHE-NRFANRHFSA-N
fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine
2756114
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
109425-55-0
C25H30N2O6
454.523
MFCD00065668
JOOIZTMAHNLNHE-NRFANRHFSA-N
fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine
2756114
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 111°C to 115°C
Beilstein 4772025
Formula Weight 454.52
Percent Purity 96%
Quantity 5 g
Chemical Name or Material Ndelta-Boc-Nalpha-Fmoc-L-ornithine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'