missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine dicyclohexylammonium salt, 95%

Brand:  Alfa Aesar H62206.03

Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine dicyclohexylammonium salt, 95%

Code : NEW

Additional Details : CAS Number : 13665-13-9 Weight : 0.00100kg

Product Code. 15487156

  • £10.28 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket