missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride, 98%, Thermo Scientific™

Catalog Number 15422599
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
5874-75-9
C14H21ClN2O4
316.78
MFCD00077027
ZUJMKNWYEVTCNT-UHFFFAOYNA-N
h-orn z-ome.hcl, h-orn z-ome hcl, h-orn z-ome-hcl, h-orn z-ome hydrochloride, c14h20n2o4.hcl, n-delta-z-l-ornithine methyl ester hydrochloride, h-orn cbz-ome inverted exclamation mark currencyhcl, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride
13967650
Cl.COC(=O)C(N)CCCNC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
5874-75-9
C14H21ClN2O4
316.78
MFCD00077027
ZUJMKNWYEVTCNT-UHFFFAOYNA-N
h-orn z-ome.hcl, h-orn z-ome hcl, h-orn z-ome-hcl, h-orn z-ome hydrochloride, c14h20n2o4.hcl, n-delta-z-l-ornithine methyl ester hydrochloride, h-orn cbz-ome inverted exclamation mark currencyhcl, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride
13967650
Cl.COC(=O)C(N)CCCNC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 141°C
Beilstein 4341483
IUPAC Name methyl 2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoate hydrochloride
Formula Weight 316.78
Percent Purity 98%
Quantity 25 g
Physical Form Solid
Chemical Name or Material Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'