missing translation for 'onlineSavingsMsg'
Learn More

NCS-382, Tocris Bioscience™

Antagonist of γ-hydroxybutyric acid

Brand:  Tocris (Bio-Techne) 078050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467993

  • £295.47 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

γ-Hydroxybutyric acid antagonist, anticonvulsant.
Specifications

Chemical Identifiers

520505-01-5
218.252
tocris-0780, 2e-2-5s-5-hydroxy-5,7,8,9-tetrahydrobenzo 7 annulen-6-ylidene acetic acid, s,e-2-5-hydroxy-8,9-dihydro-5h-benzo 7 annulen-6 7h-ylidene acetic acid
(2E)-2-[(5S)-5-hydroxy-5,7,8,9-tetrahydrobenzo[7]annulen-6-ylidene]acetic acid
C13H14O3
UADPGHINQMWEAG-FROQITRMSA-N
6604757
C1CC2=CC=CC=C2C(C(=CC(=O)O)C1)O

Specifications

50mg
520505-01-5
tocris-0780, 2e-2-5s-5-hydroxy-5,7,8,9-tetrahydrobenzo 7 annulen-6-ylidene acetic acid, s,e-2-5-hydroxy-8,9-dihydro-5h-benzo 7 annulen-6 7h-ylidene acetic acid
C1CC2=CC=CC=C2C(C(=CC(=O)O)C1)O
218.252
218.25
NCS-382
C13H14O3
UADPGHINQMWEAG-FROQITRMSA-N
(2E)-2-[(5S)-5-hydroxy-5,7,8,9-tetrahydrobenzo[7]annulen-6-ylidene]acetic acid
6604757
>99%
Videos
Documents