missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NCOA2 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Label or Dye | FITC, Texas Red |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) |
Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
Location | 8q13.3, 8q13.3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003425
|
Abnova™
FS0013.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093415
|
Abnova™
FS0013.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) | |
8q13.3, 8q13.3 | |
NCOA2 Split FISH Probe |
FITC, Texas Red | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
NCOA2, NCOA2 |