Learn More
NBQX, Tocris Bioscience™
Potent AMPA antagonist; more selective than CNQX (Cat. No. 0190)
POA
Description
Description
Potent, selective and competitive AMPA receptor antagonist. Neuroprotective and anticonvulsant; active in vivo.
Disodium Salt also available.