missing translation for 'onlineSavingsMsg'
Learn More

NBQX, Tocris Bioscience™

Potent AMPA antagonist; more selective than CNQX (Cat. No. 0190)

Brand:  Tocris (Bio-Techne) 0373/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13457593

  • £787.92 / 50mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective and competitive AMPA receptor antagonist. Neuroprotective and anticonvulsant; active in vivo.

Disodium Salt also available.

Specifications

Chemical Identifiers

118876-58-7
336.278
nbqx, 2,3-dioxo-6-nitro-7-sulfamoylbenzo f quinoxaline, 2,3-dihydroxy-6-nitro-7-sulfamoyl-benzo f quinoxaline, 6-nitro-7-sulfamoylbenzo f quinoxaline-2,3-dione, 6-nitro-2,3-dioxo-1,4-dihydrobenzo f quinoxaline-7-sulfonamide, 1,2,3,4-tetrahydro-6-nitro-2,3-dioxobenzo f quinoxaline-8-sulfonamide, 6-nitro-7-sulphamoylbenzo f quinoxaline-2,3-dione, 2,3-dihydroxy-6-nitro-7-sulfamoylbenzo f quinoxaline, 2,3-dihydroxy-6-nitro-7-sulphamoylbenzo f-quinoxaline
CHEBI:31073
C1=CC2=C3C(=CC(=C2C(=C1)S(=O)(=O)N)[N+](=O)[O-])NC(=O)C(=O)N3
C12H8N4O6S
UQNAFPHGVPVTAL-UHFFFAOYSA-N
3272524
6-nitro-2,3-dioxo-1,4-dihydrobenzo[f]quinoxaline-7-sulfonamide

Specifications

50mg
118876-58-7
nbqx, 2,3-dioxo-6-nitro-7-sulfamoylbenzo f quinoxaline, 2,3-dihydroxy-6-nitro-7-sulfamoyl-benzo f quinoxaline, 6-nitro-7-sulfamoylbenzo f quinoxaline-2,3-dione, 6-nitro-2,3-dioxo-1,4-dihydrobenzo f quinoxaline-7-sulfonamide, 1,2,3,4-tetrahydro-6-nitro-2,3-dioxobenzo f quinoxaline-8-sulfonamide, 6-nitro-7-sulphamoylbenzo f quinoxaline-2,3-dione, 2,3-dihydroxy-6-nitro-7-sulfamoylbenzo f quinoxaline, 2,3-dihydroxy-6-nitro-7-sulphamoylbenzo f-quinoxaline
C1=CC2=C3C(=CC(=C2C(=C1)S(=O)(=O)N)[N+](=O)[O-])NC(=O)C(=O)N3
336.278
CHEBI:31073
>98%
NBQX
C12H8N4O6S
UQNAFPHGVPVTAL-UHFFFAOYSA-N
6-nitro-2,3-dioxo-1,4-dihydrobenzo[f]quinoxaline-7-sulfonamide
3272524
336.28
Videos
Documents