Learn More
Tocris Bioscience™ NBQX disodium salt
Potent AMPA antagonist; more water soluble form of NBQX (Cat. No. 0373)
Brand:  Tocris Bioscience™ 1044/50
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13438373
  • £936.84 / 50mg
Estimated Shipment
14-09-2020
Description
Description
Potent, selective and competitive AMPA receptor antagonist. Neuroprotective and anticonvulsant; active in vivo. More water soluble disodium salt of NBQX (Cat. No. 0373).
Specifications
Specifications
2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide disodium salt
>98%
Observed MW: 380.24
Soluble to 100mM in water
479347-86-9
C12H6N4O6SNa2