missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Vial Crushers

Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.

Brand:  Thermo Scientific™ C4040-PVCM

Code : 17

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10129094

  • £14285.00 / Each


Estimated Shipment: 06-01-2022
Add to basket

Description

Description

Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.

  • Breaks and separates liquid from all 1mL to 100mL vials and ampules
  • Capable of processing approximately 5000 2mL vials per hour
  • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
  • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
Specifications

Specifications

Pneumatic Benchtop Vial Crusher with Stainless-steel Mobile Cart
National™
Videos
SDS
Documents
Certificates