Learn More
Thermo Scientific™ National Silanized Target™ Screw-Thread Vials
Thermo Scientific™ National Silanized Target Screw-Thread Vials provide full-surface coverage with noncontaminating, highly resistive silane reagent coating.
Brand: Thermo Scientific™ C4013-S2
Code : 50
Additional Details : Weight : 0.25000kg
Description
Thermo Scientific™ National Silanized Target Screw-Thread Vials provide full-surface coverage with noncontaminating, highly resistive silane reagent coating.
- Coating deactivates active sites of glass to prevent analyte loss
- Excellent for use with analysis of polar compounds, biological, and pH sensitive samples
- Maximum fill volume 2mL
- 12 x 32mm
Specifications
Amber | |
Glass | |
32 mm |
12 mm | |
2 mL | |
I-D |