Learn More
National Diagnostics™ Calci-Clear™ Decalcifying Solution
A solution of efficient chelating and sequestering agents used with tissue samples to remove and bind all calcium and other metal ions
Brand:  National Diagnostics™ HS-104-1x1gal
Code : LF
Additional Details : CAS Number : 64-18-6 Weight : 4.00000kg Transport : UN number : 3265 Chem class : 1a Pack group : II
 View more versions of this product
Product Code. 12984900
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Decalcifying Solution
1 gal.
Videos