missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Narrow Neck Unground Reagent Bottle

Brand:  Duran™ 211613605

Code : A0

Additional Details : Weight : 1.65000kg

 View more versions of this product

Product Code. 11777961

  • £138.00 / Pack of 10


Estimated Shipment
22-06-2018
Add to basket

Specifications

Specifications

250mL
Clear
Bottle, Reagent
70mm
Narrow
DIN 12035/ISO 4796
10Pack
Yes
133mm
Videos