missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Narrow Neck Soda Glass Reagent Bottles
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Closure Material:
Amber Glass
Clear Glass
Description
Interchangeable hollow ground amber glass stopper.
Specifications
Closure Material | Clear Glass |
Product Type | Reagent Bottle |
Capacity (Metric) | 100 mL |
Includes | Interchangeable hollow ground clear glass stopper |
Material | Soda Lime Glass |
No. per Pack | 10 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'