missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Narrow Neck Flask

Brand:  Duran™ 217217109

Code : A0

Additional Details : Weight : 0.95000kg

 View more versions of this product

Product Code. 10707571

  • £91.90 / Each


Estimated Shipment
12-02-2018
Add to basket

Specifications

Specifications

Round
207mm
Ideal for heating applications
Narrow
ISO 1773, USP Standard
4000mL
290mm
Videos