missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Narrow Neck Clear Borosilicate Glass Reagent Bottles, 250mL

Complies with ISO/EN recommendations. Poulten Graf™ Narrow Neck Borosilicate Glass Reagent Bottles have pre-printed hazard symbols and phrases where required and feature a narrow neck design.

Brand:  Poulten Graf™ SL921/D/L26

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12367238

  • £38.10 / Pack of 2


Estimated Shipment
23-02-2018
Add to basket

Description

Description

  • Interchangeable polypropylene hexagonal head stoppers
  • 250mL capacity with PVC coating
  • Can be dry heat sterilized at 140°C
Specifications

Specifications

Sulfuric acid dilute
2
Bottle, Reagent
Interchangeable Polypropylene hexagonal head stoppers
Dry heat sterilized
Videos