Poulten Graf™ Narrow Neck Clear Borosilicate Glass Reagent Bottles, 250mL

Complies with ISO/EN recommendations. Poulten Graf™ Narrow Neck Borosilicate Glass Reagent Bottles have pre-printed hazard symbols and phrases where required and feature a narrow neck design.

Overview
Brand: Poulten Graf™

Manufacturer Part Number: SL921/D/L11

Bottle labelled reagent glass hydrochloric aciddiluted clear 250mL Volac, 2/Cs, MA: TBD

UNSPSC: 24122003

Code: 02

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12317228

Quantity Price
1 £ 38.0 / Pack of 2
Estimated Shipment
03-03-2017
Add to basket


Description and Specification

Specification

No. per Pack 2
Sterility Dry heat sterilized
With Interchangeable Polypropylene hexagonal head stoppers
Product Type Reagent Bottle
Chemical Label Hydrochloric acid dilute
Mouth Narrow
Color Clear
Material Borosilicate Glass
Includes Interchangeable Polypropylene hexagonal head stoppers
Capacity (Metric) 250mL

  • Interchangeable polypropylene hexagonal head stoppers
  • 250mL capacity with PVC coating
  • Can be dry heat sterilized at 140°C

Documentation

Technical documents