missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Narrow Neck Clear Borosilicate Glass Reagent Bottles, 250mL
Click to view available options
Chemical Label:
Ammonia (Concentrated)
Codium Hydroxide (Concentrated)
Hydrochloric Acid (Concentrated)
Hydrochloric Acid (Dilute)
Nitric Acid (Dilute)
Sulfuric Acid (Concentrated)
Sulfuric Acid (Dilute)
Description
- Interchangeable polypropylene hexagonal head stoppers
- 250mL capacity with PVC coating
- Can be dry heat sterilized at 140°C
Specifications
Chemical Label | Hydrochloric Acid (Concentrated) |
Product Type | Reagent Bottle |
Includes | Interchangeable Polypropylene hexagonal head stoppers |
Material | Borosilicate Glass |
No. per Pack | 2 |
Sterility | Dry Heat Sterilized |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'