missing translation for 'onlineSavingsMsg'
Learn More

Naphthol Yellow S, Thermo Scientific™

Catalog Number p-7025415
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
846-70-8
C10H4N2Na2O8S
358.188
MFCD00003958
CTIQLGJVGNGFEW-UHFFFAOYSA-L
C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt
2724063
CHEBI:87219
disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]
This item is not returnable. View return policy
846-70-8
C10H4N2Na2O8S
358.188
MFCD00003958
CTIQLGJVGNGFEW-UHFFFAOYSA-L
C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt
2724063
CHEBI:87219
disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Beilstein 2708476
Merck Index 14,6393
Formula Weight 358.2
Quantity 50 g
Chemical Name or Material Naphthol Yellow S
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'