Learn More
Macherey-Nagel™ Nano Durasil-20 TLC Plate
Brand:  Macherey-Nagel™ 812011
Code : 50
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10504922
  • £372.00 / Pack of 50
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Specifications
Nano Durasil-20
60 Å
TLC Plate
∼500 m2/g
10 x 20 cm
2 to 10 μm
50 Pk.
0.75 mL/g
0.2 mm