Learn More
Macherey-Nagel™ Nanaocolor™ Tube Test
Perform easy, fast measurements. Macherey-Nagel™ Nanocolor™ Tube Tests are bar coded for automatic recognition by Macherey-Nagel photometers. Each test includes exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high user safety and maximum precision.
£101.00
Specifications
Certifications/Compliance | DIN, EN, ISO, EPA and APHA standards |
---|---|
Format | Pre-dosed Cuvettes |
For Use With (Application) | Photometric determination |
For Use With (Equipment) | Compatible Macherey-Nagel photometers |
Product Code | Brand | Analytes | Range | Test Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Analytes | Range | Test Method | Price | Quantity & Availability | |||
10671553
|
Macherey-Nagel™
985097 |
Tin (Sn) | 0.10 - 3.00mg/L Sn | 9-phenyl-3-fluorone |
£101.00
1 test |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Nanocolor™ tube tests for photometric analysis feature easy handling, making them the first choice for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information about the test at one glance. A variety of test kits are available with different measurement ranges for all typical parameters relevant in water and waste water analysis.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
DIN, EN, ISO, EPA and APHA standards | |
Pre-dosed Cuvettes | |
Photometric determination | |
Compatible Macherey-Nagel photometers |