Learn More
Macherey-Nagel™ Nanaocolor™ Tube Test
Perform easy, fast measurements. Macherey-Nagel™ Nanocolor™ Tube Tests are bar coded for automatic recognition by Macherey-Nagel photometers. Each test includes exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high user safety and maximum precision.
Brand: Macherey-Nagel™ 985859
152.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Nanocolor™ tube tests for photometric analysis feature easy handling, making them the first choice for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information about the test at one glance. A variety of test kits are available with different measurement ranges for all typical parameters relevant in water and waste water analysis.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
Methanol (MeOH) | |
Light pink label | |
Photometric determination | |
23 test tubes Methanol 15 R0, 1 flask with 90mL Methanol 15 R1, 1 flask with 6mL Methanol 15 R2, 1 flask with 10mL Methanol 15 R3 | |
23 Pack | |
1.5 mL | |
Alcoholoxidase / Peroxidase | |
Cuvette Test |
DIN, EN, ISO, EPA and APHA standards | |
Pre-dosed Cuvettes | |
Compatible Macherey-Nagel photometers | |
23 | |
0.2 - 15.0mg/L MeOH | |
Frozen (<0°C) | |
30 + 10 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.