missing translation for 'onlineSavingsMsg'
Learn More

NAN-190 hydrobromide, Tocris Bioscience™

5-HT1A antagonist

Brand:  Tocris (Bio-Techne) 0553250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 250mg

 View more versions of this product

Product Code. 13457753

  • £416.01 / 250mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

5-HT1A antagonist.
Specifications

Chemical Identifiers

115338-32-4
474.399
nan-190 hydrobromide, 2-4-4-2-methoxyphenyl piperazin-1-yl butyl isoindoline-1,3-dione hydrobromide, nan 190 hydrobromide, 1-2-methoxyphenyl-4-4-2-phthalimido butyl piperazine hydrobromide, 1-2-methoxyphenyl-4-4-phthalimidobutyl piperazinehydrobromide, 1-2-methoxyphenyl-4-4-phthalimidobutyl piperazine hydrobromide, 2-4-4-2-methoxyphenyl piperazin-1-yl butyl isoindole-1,3-dione hydrobromide, 2-4-4-2-methoxyphenyl piperazin-1-yl butyl-1h-isoindole-1,3 2h-dione hydrobromide
CHEBI:64123
COC1=CC=CC=C1N2CCN(CC2)CCCCN3C(=O)C4=CC=CC=C4C3=O.Br
C23H28BrN3O3
AXRUEPFPTQYHQD-UHFFFAOYSA-N
107966
2-[4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl]isoindole-1,3-dione;hydrobromide

Specifications

NAN-190 hydrobromide
115338-32-4
nan-190 hydrobromide, 2-4-4-2-methoxyphenyl piperazin-1-yl butyl isoindoline-1,3-dione hydrobromide, nan 190 hydrobromide, 1-2-methoxyphenyl-4-4-2-phthalimido butyl piperazine hydrobromide, 1-2-methoxyphenyl-4-4-phthalimidobutyl piperazinehydrobromide, 1-2-methoxyphenyl-4-4-phthalimidobutyl piperazine hydrobromide, 2-4-4-2-methoxyphenyl piperazin-1-yl butyl isoindole-1,3-dione hydrobromide, 2-4-4-2-methoxyphenyl piperazin-1-yl butyl-1h-isoindole-1,3 2h-dione hydrobromide
COC1=CC=CC=C1N2CCN(CC2)CCCCN3C(=O)C4=CC=CC=C4C3=O.Br
474.399
CHEBI:64123
>98%
250mg
C23H28BrN3O3
AXRUEPFPTQYHQD-UHFFFAOYSA-N
2-[4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl]isoindole-1,3-dione;hydrobromide
107966
474.4
Videos
Documents