missing translation for 'onlineSavingsMsg'
Learn More

Naltrindole hydrochloride, Tocris Bioscience™

Selective non-peptide δ antagonist

Brand:  Tocris (Bio-Techne) 0740/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13457933

  • £474.52 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Highly selective non-peptide δ opioid antagonist, showing 223- and 346-fold greater activity at δ than at μ? and κ opioid receptors.
Specifications

Chemical Identifiers

111469-81-9
450.963
naltrindole hydrochloride, nti hydrochloride, naltrindole hydrochloride, naltrindole hcl, naltridole hydrochloride, naltrindole hydrochloride, solid, 4,8-methanobenzofuro 2,3-a pyrido 4,3-b carbazole-1,8a 9h-diol,7-cyclopropylmethyl-5,6,7,8,14,14b-hexahydro-,hydrochloride 1:1 , 4bs,8r,8as,14br
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6.Cl
C26H27ClN2O3
KNJKRQXCFJCQHC-IDRHMUJXSA-N
24840086

Specifications

Naltrindole hydrochloride
111469-81-9
naltrindole hydrochloride, nti hydrochloride, naltrindole hydrochloride, naltrindole hcl, naltridole hydrochloride, naltrindole hydrochloride, solid, 4,8-methanobenzofuro 2,3-a pyrido 4,3-b carbazole-1,8a 9h-diol,7-cyclopropylmethyl-5,6,7,8,14,14b-hexahydro-,hydrochloride 1:1 , 4bs,8r,8as,14br
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6.Cl
24840086
>99%
50mg
C26H27ClN2O3
KNJKRQXCFJCQHC-IDRHMUJXSA-N
450.963
450.96
Videos