missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-p-Toluenesulfonyl-L-arginine, 98%, Thermo Scientific™

Catalog Number 15479908
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1159-15-5
C13H20N4O4S
328.39
MFCD00019732
KFNRNFXZFIRNEO-UHFFFAOYNA-N
tos-arg-oh, n-tosyl-l-arginine, n-p-tosyl-l-arginine, tosyl-l-arginine, nalpha-tosyl-l-arginine, nalpha-p-toluenesulfonyl-l-arginine, p-tosyl-l-arginine, l-arginine, n2-4-methylphenyl sulfonyl, 2s-5-carbamimidamido-2-4-methylbenzenesulfonamido pentanoic acid, tosyl arginine
52501
(2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(O)=O
This item is not returnable. View return policy
1159-15-5
C13H20N4O4S
328.39
MFCD00019732
KFNRNFXZFIRNEO-UHFFFAOYNA-N
tos-arg-oh, n-tosyl-l-arginine, n-p-tosyl-l-arginine, tosyl-l-arginine, nalpha-tosyl-l-arginine, nalpha-p-toluenesulfonyl-l-arginine, p-tosyl-l-arginine, l-arginine, n2-4-methylphenyl sulfonyl, 2s-5-carbamimidamido-2-4-methylbenzenesulfonamido pentanoic acid, tosyl arginine
52501
(2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 328.39
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material Nalpha-p-Toluenesulfonyl-L-arginine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'