missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine, 95%

Brand:  Alfa Aesar™ H62243.14

Code : NEW

Additional Details : CAS Number : 76265-69-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15438798

  • £262.80 / 25g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine
464.43
Fmoc-Lys(Tfa)-OH
76265-69-5
MFCD00153360

Specifications

Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine
Solid
464.43
Fmoc-Lys(Tfa)-OH
25g
76265-69-5
95%
MFCD00153360
170° to 174°C
Videos
SDS