missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine, 95%, Thermo Scientific Chemicals

Catalog Number 15400429
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
167393-62-6
C41H40N2O4
624.78
MFCD00237166
YPTNAIDIXCOZAJ-WHUAGQPENA-N
fmoc-lys mtt-oh, fmoc-l-lys mtt-oh, fmoc-n'-methyltrityl-l-lysine, nalpha-fmoc-nepsilon-4-methyltrityl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, fmoc-lys meotrt-oh, fmoc-lysine mtt-oh, fmoc-l-lys mmt-oh, ambotzfaa1135, fmoc-lys ch3trt-oh
10963195
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[[(4-methylphenyl)-diphenylmethyl]amino]hexanoic acid
CC1=CC=C(C=C1)C(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
167393-62-6
C41H40N2O4
624.78
MFCD00237166
YPTNAIDIXCOZAJ-WHUAGQPENA-N
fmoc-lys mtt-oh, fmoc-l-lys mtt-oh, fmoc-n'-methyltrityl-l-lysine, nalpha-fmoc-nepsilon-4-methyltrityl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, fmoc-lys meotrt-oh, fmoc-lysine mtt-oh, fmoc-l-lys mmt-oh, ambotzfaa1135, fmoc-lys ch3trt-oh
10963195
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[[(4-methylphenyl)-diphenylmethyl]amino]hexanoic acid
CC1=CC=C(C=C1)C(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 624.77
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'