missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine, 95%

Brand:  Alfa Aesar™ H63629.06

Code : NEW

Additional Details : CAS Number : 167393-62-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410429

  • £123.10 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
624.77
Fmoc-Lys(Mtt)-OH
167393-62-6
MFCD00237166

Specifications

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
Solid
624.77
Fmoc-Lys(Mtt)-OH
167393-62-6
95%
MFCD00237166
5g
Videos
SDS