missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Fmoc-D-asparagine, 95%

Brand:  Alfa Aesar™ H62691.14

Code : NEW

Additional Details : CAS Number : 108321-39-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15459178

  • £240.62 / 25g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Fmoc-D-asparagine
354.36
Fmoc-D-Asn-OH
108321-39-7
MFCD00062956

Specifications

Nalpha-Fmoc-D-asparagine
Solid
354.36
Fmoc-D-Asn-OH
25g
108321-39-7
95%
MFCD00062956
≈185°C (dec.)
Videos
SDS