missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Fmoc-1-Boc-L-histidine cyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H62237.14

Code : NEW

Additional Details : CAS Number : 210820-99-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15437196

  • £298.92 / 25g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Fmoc-1-Boc-L-histidine cyclohexylammonium salt
572.69
Fmoc-His(Boc)-OH.CHA
210820-99-8
MFCD00153357

Specifications

Nalpha-Fmoc-1-Boc-L-histidine cyclohexylammonium salt
Solid
572.69
Fmoc-His(Boc)-OH.CHA
210820-99-8
95%
MFCD00153357
25g
Videos
SDS