missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Fmoc-1-Boc-L-histidine cyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H62237.06

Code : NEW

Additional Details : CAS Number : 210820-99-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15427196

  • £59.78 / 5g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Fmoc-1-Boc-L-histidine cyclohexylammonium salt
572.69
Fmoc-His(Boc)-OH.CHA
210820-99-8
MFCD00153357

Specifications

Nalpha-Fmoc-1-Boc-L-histidine cyclohexylammonium salt
Solid
572.69
Fmoc-His(Boc)-OH.CHA
210820-99-8
95%
MFCD00153357
5g
Videos
SDS