missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nomega-(p-toluenesulfonyl)-L-arginine, 95%

Brand:  Alfa Aesar™ H62953.06

Code : NEW

Additional Details : CAS Number : 13836-37-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449398

  • £29.15 / 5g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Nomega-(p-toluenesulfonyl)-L-arginine
428.5
2683538
13836-37-8
MFCD00065557
Boc-Arg(Tos)-OH; Nα-Boc-Nω-tosyl-L-arginine

Specifications

Nalpha-Boc-Nomega-(p-toluenesulfonyl)-L-arginine
Solid
428.5
2683538
≈90°C
13836-37-8
95%
MFCD00065557
Boc-Arg(Tos)-OH; Nα-Boc-Nω-tosyl-L-arginine
5g
Videos
SDS