missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nomega-nitro-D-arginine, 98%

Brand:  Alfa Aesar™ H62885.03

Code : NEW

Additional Details : CAS Number : 50913-12-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419338

  • £12.30 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Nomega-nitro-D-arginine
319.31
Boc-D-Arg(NO2)-OH; (R)-2-(Boc-amino)-5-(3-nitroguanidino)pentanoic acid
50913-12-7
MFCD00038091

Specifications

Nalpha-Boc-Nomega-nitro-D-arginine
Solid
319.31
Boc-D-Arg(NO2)-OH; (R)-2-(Boc-amino)-5-(3-nitroguanidino)pentanoic acid
50913-12-7
98%
MFCD00038091
1g
Videos
SDS