missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nomega-mesitylenesulfonyl-L-arginine cyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H63957.03

Code : NEW

Additional Details : CAS Number : 68262-71-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418606

  • £29.89 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Nomega-mesitylenesulfonyl-L-arginine cyclohexylammonium salt
555.73
Boc-Arg(Mts)-OH.CHA
68262-71-5
MFCD00236820

Specifications

Nalpha-Boc-Nomega-mesitylenesulfonyl-L-arginine cyclohexylammonium salt
Solid
555.73
Boc-Arg(Mts)-OH.CHA
1g
68262-71-5
98%
MFCD00236820
130°C
Videos
SDS