missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 98%

Brand:  Alfa Aesar™ H62515.03

Code : NEW

Additional Details : CAS Number : 102185-38-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437516

  • £9.90 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine
486.53
Boc-Arg(Mtr)-OH
102185-38-6
MFCD00043097

Specifications

Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine
Powder
486.53
Boc-Arg(Mtr)-OH
1g
102185-38-6
98%
MFCD00043097
White to off-ehite
Videos
SDS