missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine, 95%

Brand:  Alfa Aesar™ H62873.03

Code : NEW

Additional Details : CAS Number : 200122-49-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407946

  • £45.90 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine
486.58
Boc-D-Arg(Mtr)-OH
200122-49-2
MFCD00151863

Specifications

Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine
Powder
486.58
Boc-D-Arg(Mtr)-OH
1g
200122-49-2
95%
MFCD00151863
White
Videos
SDS