missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Ngamma-xanthyl-L-asparagine, 98%, Thermo Scientific™

Catalog Number p-7055176
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
65420-40-8
C22H24N2O6
412.44
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
7017974
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
65420-40-8
C22H24N2O6
412.44
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
7017974
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 177°C to 181°C (decomposition)
Color Orange
Beilstein 5172403
Formula Weight 412.44
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Ngamma-xanthyl-L-asparagine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'